Anti-Neomycin Mouse monoclonal antibody [clone: 1.BB.912]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15068-100EA 4250 DKK
ABCAAB15068-100
Anti-Neomycin Mouse monoclonal antibody [clone: 1.BB.912]
Type: Primary
Antigen: Neomycin
Clonality: monoclonal
Clone: 1.BB.912
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR