Anti-Amoxicillin Mouse monoclonal antibody [clone: 1.BB.832]
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB15054-100EA 4250 DKK
ABCAAB15054-100
Anti-Amoxicillin Mouse monoclonal antibody [clone: 1.BB.832]
Type: Primary
Antigen: Amoxicillin
Clonality: monoclonal
Clone: 1.BB.832
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu