Anti-CEBPB Mouse monoclonal antibody unconjugated [clone: 18F8]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15049-100EA 5810 DKK
ABCAAB15049-100
Anti-CEBPB Mouse monoclonal antibody unconjugated [clone: 18F8]
Antistoffer
Type: Primary
Antigen: CEBPB (CCAAT/enhancer binding protein (C/EBP), beta)
Clonality: monoclonal
Clone: 18F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR