Anti-RPS6 Rabbit monoclonal antibody [clone: 4H372]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB15031-50EA 0 DKK
ABCAAB15031-50
Anti-RPS6 Rabbit monoclonal antibody [clone: 4H372]
Type: Primary
Antigen: RPS6 (ribosomal protein S6)
Clonality: monoclonal
Clone: 4H372
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR