Anti-EFEMP1 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14926-50EA 5200 DKK
ABCAAB14926-50
Anti-EFEMP1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: EFEMP1 (EGF containing fibulin-like extracellular matrix protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR