Anti-SIGLECL1 Goat polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14752-100EA 2820 DKK
ABCAAB14752-100
Anti-SIGLECL1 Goat polyclonal antibody
Type: Primary
Antigen: SIGLECL1 (SIGLEC family like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR