Anti-cardiac FABP Mouse monoclonal antibody [clone: 6B6]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14727-200EA 4300 DKK
ABCAAB14727-200
Anti-cardiac FABP Mouse monoclonal antibody [clone: 6B6]
Type: Primary
Antigen: cardiac FABP
Clonality: monoclonal
Clone: 6B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR