Anti-SDHA Mouse Monoclonal Antibody [clone: 2E3GC12FB2AE2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14715-100EA 3917.53 DKK
ABCAAB14715-100
Anti-SDHA Mouse Monoclonal Antibody [clone: 2E3GC12FB2AE2]
Antistoffer
Type: Primary
Antigen: SDHA (succinate dehydrogenase complex, subunit A, flavoprotein (Fp))
Clonality: monoclonal
Clone: 2E3GC12FB2AE2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR