Anti-BNP Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 24C5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14699-200EA 5330 DKK
ABCAAB14699-200
Anti-BNP Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 24C5]
Antistoffer
Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: monoclonal
Clone: 24C5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR