Anti-PDE1C Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14602-100EA 0 DKK
ABCAAB14602-100
Anti-PDE1C Rabbit polyclonal antibody
Type: Primary
Antigen: PDE1C (phosphodiesterase 1C, calmodulin-dependent 70kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR