Anti-GDF15 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14586-50EA 0 DKK
ABCAAB14586-50
Anti-GDF15 Rabbit polyclonal antibody
Type: Primary
Antigen: GDF15 (growth differentiation factor 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR