Anti-LHX3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14555-50EA 5450 DKK
ABCAAB14555-50
Anti-LHX3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LHX3 (LIM homeobox 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR