Anti-PTH Mouse Monoclonal Antibody [clone: D1.1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14498-100EA 5670 DKK
ABCAAB14498-100
Anti-PTH Mouse Monoclonal Antibody [clone: D1.1]
Antistoffer
Type: Primary
Antigen: PTH (parathyroid hormone)
Clonality: monoclonal
Clone: D1.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR