Anti-GCDFP 15 Mouse monoclonal antibody [clone: 3G153]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14479-100EA 4300 DKK
ABCAAB14479-100
Anti-GCDFP 15 Mouse monoclonal antibody [clone: 3G153]
Type: Primary
Antigen: GCDFP 15
Clonality: monoclonal
Clone: 3G153
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR