Anti-SUPT16H Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14466-50EA 4250 DKK
ABCAAB14466-50
Anti-SUPT16H Rabbit polyclonal antibody
Type: Primary
Antigen: SUPT16H (suppressor of Ty 16 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR