Anti-CCKBR Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14440-50EA 5760 DKK
ABCAAB14440-50
Anti-CCKBR Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CCKBR (cholecystokinin B receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR