Anti-BID Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14435-100EA 4690 DKK
ABCAAB14435-100
Anti-BID Rabbit polyclonal antibody
Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR