Anti-ELAVL4 Mouse monoclonal antibody [clone: 16C12]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14369-100EA 4060 DKK
ABCAAB14369-100
Anti-ELAVL4 Mouse monoclonal antibody [clone: 16C12]
Type: Primary
Antigen: ELAVL4 (ELAV like neuron-specific RNA binding protein 4)
Clonality: monoclonal
Clone: 16C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR