Anti-Thymosin beta 4 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB14334-50EA 3949.41 DKK
ABCAAB14334-50
Anti-Thymosin beta 4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Thymosin beta 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human