Anti-RGS8 Chicken Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14267-50EA 5820 DKK
ABCAAB14267-50
Anti-RGS8 Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RGS8 (regulator of G-protein signaling 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR