Anti-RGS14 Chicken polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14262-50EA 4670 DKK
ABCAAB14262-50
Anti-RGS14 Chicken polyclonal antibody
Type: Primary
Antigen: RGS14 (regulator of G-protein signaling 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR