Anti-POLR2K Chicken Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14253-50EA 6150 DKK
ABCAAB14253-50
Anti-POLR2K Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: POLR2K (polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR