Anti-D4 GDI Chicken polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14230-50EA 4600 DKK
ABCAAB14230-50
Anti-D4 GDI Chicken polyclonal antibody
Type: Primary
Antigen: D4 GDI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR