Anti-AP3B2 Chicken Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14227-50EA 5880 DKK
ABCAAB14227-50
Anti-AP3B2 Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AP3B2 (adaptor-related protein complex 3, beta 2 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR