Anti-CEA Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14191-50EA 4250 DKK
ABCAAB14191-50
Anti-CEA Rabbit polyclonal antibody
Type: Primary
Antigen: CEA (Carcino Embryonic Antigen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR