Anti-SPARC Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14174-50EA 4284.74 DKK
ABCAAB14174-50
Anti-SPARC Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SPARC (secreted protein, acidic, cysteine-rich (osteonectin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR