Anti-ACTB Mouse Monoclonal Antibody [clone: 2Q1055]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14128-50EA 5620 DKK
ABCAAB14128-50
Anti-ACTB Mouse Monoclonal Antibody [clone: 2Q1055]
Antistoffer
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone: 2Q1055
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR