Anti-14-3-3 sigma Mouse monoclonal antibody [clone: 1.N.6]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB14116-50EA 4300 DKK
ABCAAB14116-50
Anti-14-3-3 sigma Mouse monoclonal antibody [clone: 1.N.6]
Type: Primary
Antigen: 14-3-3 sigma (14-3-3 sigma)
Clonality: monoclonal
Clone: 1.N.6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR