Anti-FIV p120 Mouse monoclonal antibody [clone: vpg 68]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13969-1EA 4140 DKK
ABCAAB13969-1
Anti-FIV p120 Mouse monoclonal antibody [clone: vpg 68]
Type: Primary
Antigen: FIV p120
Clonality: monoclonal
Clone: vpg 68
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR