Anti-HTR1B Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13896-100EA 5880 DKK
ABCAAB13896-100
Anti-HTR1B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HTR1B (5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR