Anti-BCL2L1 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13891-100EA 4690 DKK
ABCAAB13891-100
Anti-BCL2L1 Rabbit polyclonal antibody
Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR