Anti-SH3GLB1 Mouse monoclonal antibody [clone: 30A882]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13886-100EA 4060 DKK
ABCAAB13886-100
Anti-SH3GLB1 Mouse monoclonal antibody [clone: 30A882]
Type: Primary
Antigen: SH3GLB1 (SH3-domain GRB2-like endophilin B1)
Clonality: monoclonal
Clone: 30A882
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR