Anti-ADIPOQ Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13881-50EA 5430 DKK
ABCAAB13881-50
Anti-ADIPOQ Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR