Anti-ANTXR1 Mouse Monoclonal Antibody [clone: 200C1339]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13798-100EA 5810 DKK
ABCAAB13798-100
Anti-ANTXR1 Mouse Monoclonal Antibody [clone: 200C1339]
Antistoffer
Type: Primary
Antigen: ANTXR1 (anthrax toxin receptor 1)
Clonality: monoclonal
Clone: 200C1339
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR