Anti-IL1B Rat monoclonal antibody [clone: 1400.24.17]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13784-100EA 4560 DKK
ABCAAB13784-100
Anti-IL1B Rat monoclonal antibody [clone: 1400.24.17]
Type: Primary
Antigen: IL1B (interleukin 1, beta)
Clonality: monoclonal
Clone: 1400.24.17
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR