Anti-RHOBTB2 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13738-200EA 4250 DKK
ABCAAB13738-200
Anti-RHOBTB2 Rabbit polyclonal antibody
Type: Primary
Antigen: RHOBTB2 (Rho-related BTB domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR