Anti-BCL3 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13708-200EA 4060 DKK
ABCAAB13708-200
Anti-BCL3 Rabbit polyclonal antibody
Type: Primary
Antigen: BCL3 (B-cell CLL/lymphoma 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR