Anti-LCK Mouse monoclonal antibody [clone: 33D196]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13699-100EA 4060 DKK
ABCAAB13699-100
Anti-LCK Mouse monoclonal antibody [clone: 33D196]
Type: Primary
Antigen: LCK (lymphocyte-specific protein tyrosine kinase)
Clonality: monoclonal
Clone: 33D196
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR