Anti-PMAIP1 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13687-100EA 4140 DKK
ABCAAB13687-100
Anti-PMAIP1 Rabbit polyclonal antibody
Type: Primary
Antigen: PMAIP1 (phorbol-12-myristate-13-acetate-induced protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR