Anti-CLEC2D Mouse monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13664-100EA 4430 DKK
ABCAAB13664-100
Anti-CLEC2D Mouse monoclonal antibody
Type: Primary
Antigen: CLEC2D (C-type lectin domain family 2, member D)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR