Anti-PMAIP1 Mouse Monoclonal Antibody [clone: 114C307]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13654-100EA 4439.87 DKK
ABCAAB13654-100
Anti-PMAIP1 Mouse Monoclonal Antibody [clone: 114C307]
Antistoffer
Type: Primary
Antigen: PMAIP1 (phorbol-12-myristate-13-acetate-induced protein 1)
Clonality: monoclonal
Clone: 114C307
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR