Anti-SOD2 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13534-50EA 6040 DKK
ABCAAB13534-50
Anti-SOD2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SOD2 (superoxide dismutase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR