Anti-LAMP2 Rat Monoclonal Antibody [clone: GL2A7]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13524-100EA 6040 DKK
ABCAAB13524-100
Anti-LAMP2 Rat Monoclonal Antibody [clone: GL2A7]
Antistoffer
Type: Primary
Antigen: LAMP2 (lysosomal-associated membrane protein 2)
Clonality: monoclonal
Clone: GL2A7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR