Anti-IL21R Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB13268-50EA 5550 DKK
ABCAAB13268-50
Anti-IL21R Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IL21R (interleukin 21 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR