Anti-Melanoma Mouse Monoclonal Antibody [clone: PNL2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB12502-250EA 3805.6 DKK
ABCAAB12502-250
Anti-Melanoma Mouse Monoclonal Antibody [clone: PNL2]
Antistoffer
Type: Primary
Antigen: Melanoma
Clonality: monoclonal
Clone: PNL2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR