Anti-RAD52 Mouse monoclonal antibody [clone: 5H9]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB12447-50EA 4650 DKK
ABCAAB12447-50
Anti-RAD52 Mouse monoclonal antibody [clone: 5H9]
Type: Primary
Antigen: RAD52 (RAD52 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: 5H9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR