Anti-MAPT Mouse Monoclonal Antibody Biotin [clone: TAU 2]
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB12357-25EA 4140 DKK
ABCAAB12357-25
Anti-MAPT Mouse Monoclonal Antibody Biotin [clone: TAU 2]
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: TAU 2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu