Anti-SCG2 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB12241-100EA 5500 DKK
ABCAAB12241-100
Anti-SCG2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SCG2 (secretogranin II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat