Anti-PLK1 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB12157-100EA 4098.45 DKK
ABCAAB12157-100
Anti-PLK1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PLK1 (polo-like kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human