Anti-Twilight Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB12131-100EA 0 DKK
ABCAAB12131-100
Anti-Twilight Rabbit Polyclonal Antibody
Type: Primary
Antigen: Twilight
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR